Order Entry
United Kingdom
ContactUsLinkComponent
Anti-BTBD10 Mouse Polyclonal Antibody
Anti-BTBD10 Mouse Polyclonal Antibody
Catalog # ABNOH00084280-B01P
Supplier:  Abnova
undefined
Anti-BTBD10 Mouse Polyclonal Antibody
Catalog # ABNOH00084280-B01P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BTB (POZ) Domain Containing 10
  • Antigen symbol:
    BTBD10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    84280
  • Antigen synonyms:
    GMRP1|BTBDA_HUMAN.|BTB domain containing 10|K+ channel tetramerization protein|GMRP 1|BTB POZ domain containing 10|BTB/POZ domain-containing protein 10|BTB broad complex tramtrack and bric a brac pox virus and zinc finger domain containing protein 10
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µg

Specifications

About this item

Type: Primary
Antigen: BTBD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human