Anti-RAB34 Mouse Monoclonal Antibody [clone: 3A5]
ABNOH00083871-M06
: Abnova
- Antibody type:Primary
- Antigen name:Ras-related protein Rab-34
- Antigen symbol:RAB34
- Clonality:Monoclonal
- Clone:3A5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:83871
- Antigen synonyms:RAB39|RAH
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: RAB34
Clonality: Monoclonal
Clone: 3A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human