Anti-ZSCAN16 Mouse Polyclonal Antibody
Catalog # ABNOH00080345-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger And SCAN Domain Containing 16
- Antigen symbol:ZSCAN16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:80345
- Antigen synonyms:ZNF435|ZNF392|dJ265C24.3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: ZSCAN16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human