Anti-FKBP10 Mouse Monoclonal Antibody [clone: 3B5]
Catalog # ABNOH00060681-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:FK506 binding protein 10
- Antigen symbol:FKBP10
- Clonality:Monoclonal
- Clone:3B5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:60681
- Antigen synonyms:Immunophilin FKBP65|FKBP65|65 kDa FK506-binding protein|FKBP10|PSEC0056|Rotamase|Peptidyl-prolyl cis-trans isomerase|FK506-binding protein 10|PPIase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: FKBP10
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human