Anti-CORO1B Mouse Monoclonal Antibody [clone: 1E7]
ABNOH00057175-M01
: Abnova
- Antibody type:Primary
- Antigen name:Coronin 1B
- Antigen symbol:CORO1B
- Clonality:Monoclonal
- Clone:1E7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:57175
- Antigen synonyms:CORONIN-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CORO1B
Clonality: Monoclonal
Clone: 1E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human