Anti-APOM Mouse Monoclonal Antibody [clone: 6H3]
ABNOH00055937-M06
: Abnova
- Antibody type:Primary
- Antigen name:Apolipoprotein M
- Antigen symbol:APOM
- Clonality:Monoclonal
- Clone:6H3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:55937
- Antigen synonyms:G3a|HSPC336|NG20 |NG20-like protein|NG20|MGC22400
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: APOM
Clonality: Monoclonal
Clone: 6H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human