Anti-IL26 Mouse Monoclonal Antibody [clone: 2A8]
Catalog # ABNOH00055801-M08
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 26
- Antigen symbol:IL26
- Clonality:Monoclonal
- Clone:2A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:55801
- Antigen synonyms:IL26|AK155|IL-26
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: IL26
Clonality: Monoclonal
Clone: 2A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human