Anti-RHOT1 Mouse Monoclonal Antibody [clone: 4H4]
Catalog # ABNOH00055288-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ras Homolog Family Member T1
- Antigen symbol:RHOT1
- Clonality:Monoclonal
- Clone:4H4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:55288
- Antigen synonyms:Ras homolog family member T1|mitochondrial Rho GTPase 1|MIRO-1|ARHT1|MIRO1|MIRO
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: RHOT1
Clonality: Monoclonal
Clone: 4H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human