Anti-RNF125 Mouse Polyclonal Antibody
Catalog # ABNOH00054941-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ring Finger Protein 125
- Antigen symbol:RNF125
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:54941
- Antigen synonyms:TRAC 1|T cell ring protein identied in activation screen|Ring finger protein 125|E3 ubiquitin protein ligase RNF125|RNF 125|T cell RING activation protein 1|RN125_HUMAN.|TRAC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RNF125
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human