Anti-APTX Mouse Polyclonal Antibody
Catalog # ABNOH00054840-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Aprataxin
- Antigen symbol:APTX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:54840
- Antigen synonyms:FHA-HIT|AOA|AXA1|EAOH|POTE ankyrin Domain Family Member G|AOA1|EOAHA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: APTX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human