Order Entry
United Kingdom
ContactUsLinkComponent
Anti-EHF Mouse Polyclonal Antibody
Anti-EHF Mouse Polyclonal Antibody
Catalog # ABNOH00026298-B01P
Supplier:  Abnova
undefined
Anti-EHF Mouse Polyclonal Antibody
Catalog # ABNOH00026298-B01P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ets homologous factor
  • Antigen symbol:
    EHF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    26298
  • Antigen synonyms:
    ESE3|ESEJ|ESE3B
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µg

Specifications

About this item

Type: Primary
Antigen: EHF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human