Anti-ARL5A Mouse Monoclonal Antibody [clone: 1F12-1A6]
ABNOH00026225-M01
: Abnova
- Antibody type:Primary
- Antigen name:ADP-Ribosylation Factor-Like 5A
- Antigen symbol:ARL5A
- Clonality:Monoclonal
- Clone:1F12-1A6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:26225
- Antigen synonyms:ARFLP5, ARL5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ARL5A
Clonality: Monoclonal
Clone: 1F12-1A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human