Anti-ZCCHC11 Mouse Polyclonal Antibody
Catalog # ABNOH00023318-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger, CCHC Domain Containing 11
- Antigen symbol:ZCCHC11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:23318
- Antigen synonyms:6030404K05Rik|Zinc finger CCHC domain containing 11|KIAA0191|PPAPD3|mKIAA0191|zinc finger CCHC domain-containing protein 11|9230115F04Rik|RP23-406B13.1|TUTase 4|terminal uridylyltransferase 4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: ZCCHC11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human