Anti-PEG10 Mouse Polyclonal Antibody
ABNOH00023089-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Paternally Expressed 10
- Antigen symbol:PEG10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:23089
- Antigen synonyms:Mart2|Mar2|HB-1|RGAG3|Edr|MEF3L|KIAA1051|PEG10
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: PEG10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human