Anti-CNOT1 Mouse Polyclonal Antibody
ABNOH00023019-A01
: Abnova
- Antibody type:Primary
- Antigen name:CCR4-NOT transcription complex, subunit 1
- Antigen symbol:CNOT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:23019
- Antigen synonyms:CDC39|NOT1H|AD-005|NOT1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: CNOT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human