Anti-PTP4A3 Mouse Polyclonal Antibody
ABNOH00011156-A01
: Abnova
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase type IVA, member 3
- Antigen symbol:PTP4A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:11156
- Antigen synonyms:PRL-R|Protein-tyrosine phosphatase 4a3|PRL-3|Protein tyrosine phosphatase type IVA protein 3|PRL3|PTP4A3|Protein-tyrosine phosphatase of regenerating liver 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: PTP4A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human