Anti-DEAF1 Mouse Polyclonal Antibody
ABNOH00010522-A01
: Abnova
- Antibody type:Primary
- Antigen name:DEAF1 transcription factor
- Antigen symbol:DEAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10522
- Antigen synonyms:MRD24|Deaf1|SPN|ZMYND5|Deformed epidermal autoregulatory factor 1 homolog|NUDR|Nuclear DEAF-1-related transcriptional regulator
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: DEAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human