Anti-CLEC10A Mouse Polyclonal Antibody
Catalog # ABNOH00010462-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:C-type lectin Domain family 10, member A
- Antigen symbol:CLEC10A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10462
- Antigen synonyms:CD301|MGL|HML|CLECSF14|HML2|CLECSF13
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: CLEC10A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human