Anti-KLRG1 Mouse Polyclonal Antibody
ABNOH00010219-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Killer cell lectin-like receptor subfamily G member 1
- Antigen symbol:KLRG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10219
- Antigen synonyms:MAFA|MAFA-2F1|2F1|CLEC15A|MAFA-LIKE|MAFA-L
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: KLRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human