Anti-ULK2 Mouse Monoclonal Antibody [clone: 6C12]
ABNOH00009706-M01
: Abnova
- Antibody type:Primary
- Antigen name:Unc-51 like autophagy activating kinase 2
- Antigen symbol:ULK2
- Clonality:Monoclonal
- Clone:6C12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9706
- Antigen synonyms:KIAA0623|ATG1B|ULK2|Unc51.2|Unc-51-like kinase 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ULK2
Clonality: Monoclonal
Clone: 6C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human