Anti-TAOK2 Mouse Monoclonal Antibody [clone: 2H6]
Catalog # ABNOH00009344-M08
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:TAO kinase 2
- Antigen symbol:TAOK2
- Clonality:Monoclonal
- Clone:2H6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9344
- Antigen synonyms:PSK1|B230344N16|PSK|MAP3K17|mKIAA0881|TAO2|TAO1|1110033K02Rik
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TAOK2
Clonality: Monoclonal
Clone: 2H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human