Anti-LRRFIP1 Mouse Monoclonal Antibody [clone: 4E11]
Catalog # ABNOH00009208-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Leucine Rich Repeat (In FLII) Interacting Protein 1
- Antigen symbol:LRRFIP1
- Clonality:Monoclonal
- Clone:4E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9208
- Antigen synonyms:LRRFIP1|LRR FLII-interacting protein 1|FLAP1|GCF2|HUFI-1|FLAP-1|FLIIAP1|GCF-2|TRIP|Leucine-rich repeat flightless-interacting protein 1|GC-binding factor 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: LRRFIP1
Clonality: Monoclonal
Clone: 4E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human