Anti-FUBP1 Mouse Monoclonal Antibody [clone: 1E10]
Catalog # ABNOH00008880-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Far upstream element binding protein 1
- Antigen symbol:FUBP1
- Clonality:Monoclonal
- Clone:1E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:8880
- Antigen synonyms:FUBP|hDH V|FBP
- Storage buffer:In 1x PBS, pH 7,4
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:FUBP1 (AAH17247.1, 1 a.a. ~ 653 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against a full-length recombinant FUBP1.
Recommended Dilutions: Western Blot (1 - 5 µg/ml)
Type: Primary
Antigen: FUBP1
Clonality: Monoclonal
Clone: 1E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human