Anti-CDC123 Mouse Monoclonal Antibody [clone: 1A1]
ABNOH00008872-M03
: Abnova
- Antibody type:Primary
- Antigen name:Cell division cycle 123
- Antigen symbol:CDC123
- Clonality:Monoclonal
- Clone:1A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8872
- Antigen synonyms:C10orf7|D123
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CDC123
Clonality: Monoclonal
Clone: 1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human