Anti-TNFRSF6B Mouse Monoclonal Antibody [clone: 7G5]
Catalog # ABNOH00008771-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Receptor Superfamily Member 6B
- Antigen symbol:TNFRSF6B
- Clonality:Monoclonal
- Clone:7G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8771
- Antigen synonyms:DJ583P15.1.1|Tumor necrosis factor receptor superfamily member 6B|TR6|DcR3|M68|UNQ186/PRO212|Decoy receptor 3|DCR3|M68E
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TNFRSF6B
Clonality: Monoclonal
Clone: 7G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human