Anti-PDLIM4 Mouse Polyclonal Antibody
ABNOH00008572-B01P
: Abnova
- Antibody type:Primary
- Antigen name:PDZ And LIM Domain 4
- Antigen symbol:PDLIM4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:8572
- Antigen synonyms:PDZ and LIM domain 4|RIL|PDLIM4|LIM domain protein|enigma homolog
- Storage buffer:In 1x PBS, pH 7,4
- Sequence:MPHSVTLRGPSPWGFRLVGGRDFSAPLTISRVHAGSKAALAALCPGDLIQAINGESTELMTHLEAQNRIKGCHDHLTLSVSRPEGRSWPSAPDDSKAQAHRIHIDPEIQDGSPTTSRRPSGTGTGPEDGRPSLGSPYGQPPRFPVPHNGSSEATLPAQMSTLHVSPPPSADPARGLPRSRDCRVDLGSEVYRMLREPAEPVAAEPKQSGSFRYLQGMLEVGEGGAPSSRHGTSSTIPSASCAVTAA
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:PDLIM4 (AAH03096, 1 a.a. ~ 246 a.a) full-length human protein.
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: PDLIM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human