Anti-PLA2G10 Rabbit Polyclonal Antibody
Catalog # ABNOH00008399-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Phospholipase A2, Group X
- Antigen symbol:PLA2G10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8399
- Antigen synonyms:SPLA2|GXPLA2|GXSPLA2|Group X Secretory Phospholipase A2|SPLA2-X|Phosphatidylcholine 2-Acylhydrolase 10
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PLA2G10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human