Anti-AKAP17A Mouse Monoclonal Antibody [clone: 2G8]
ABNOH00008227-M02
: Abnova
- Antibody type:Primary
- Antigen name:AKAP17A
- Antigen symbol:AKAP17A
- Clonality:Monoclonal
- Clone:2G8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8227
- Antigen synonyms:CXYorf3|721P|PRKA17A|DXYS155E|SFRS17A|XE7Y|CCDC133|XE7|AKAP-17A
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: AKAP17A
Clonality: Monoclonal
Clone: 2G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human