Anti-ST8SIA2 Mouse Monoclonal Antibody [clone: 6G6]
Catalog # ABNOH00008128-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:ST8 Alpha-N-Acetyl-Neuraminide Alpha-2,8-Sialyltransferase 2
- Antigen symbol:ST8SIA2
- Clonality:Monoclonal
- Clone:6G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8128
- Antigen synonyms:HsT19690|SIAT8B|ST8SIA-II|STX
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ST8SIA2
Clonality: Monoclonal
Clone: 6G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human