Anti-TPD52 Mouse Polyclonal Antibody
ABNOH00007163-A01
: Abnova
- Antibody type:Primary
- Antigen name:Tumor Protein D52
- Antigen symbol:TPD52
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7163
- Antigen synonyms:N8L|hD52|tumor protein D52|PC-1|PC1/MRPS28 fusion protein|D52|prostate and colon associated protein|TPD52|PrLZ
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: TPD52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human