Anti-TFF3 Mouse Monoclonal Antibody [clone: 3D9]
Catalog # ABNOH00007033-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Trefoil factor 3
- Antigen symbol:TFF3
- Clonality:Monoclonal
- Clone:3D9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7033
- Antigen synonyms:TFF3|ITF|TFI|hITF|Intestinal trefoil factor|P1B|Trefoil factor 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TFF3
Clonality: Monoclonal
Clone: 3D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human