Anti-SUPT5H Mouse Monoclonal Antibody [clone: 5C8]
Catalog # ABNOH00006829-M02A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Suppressor of Ty 5 Homolog
- Antigen symbol:SUPT5H
- Clonality:Monoclonal
- Clone:5C8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6829
- Antigen synonyms:Tat-CT1|SPT5|SPT5H
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: SUPT5H
Clonality: Monoclonal
Clone: 5C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat