Anti-RPS15 Mouse Monoclonal Antibody [clone: 3F8]
ABNOH00006209-M17
: Abnova
- Antibody type:Primary
- Antigen name:Ribosomal Protein S15
- Antigen symbol:RPS15
- Clonality:Monoclonal
- Clone:3F8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6209
- Antigen synonyms:RIG|S15
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: RPS15
Clonality: Monoclonal
Clone: 3F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human