Anti-KLK10 Mouse Monoclonal Antibody [clone: 1B9]
Catalog # ABNOH00005655-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Kallikrein-related peptidase 10
- Antigen symbol:KLK10
- Clonality:Monoclonal
- Clone:1B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5655
- Antigen synonyms:NES1|PRSSL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: KLK10
Clonality: Monoclonal
Clone: 1B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human