Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CNOT2 Mouse Polyclonal Antibody
Anti-CNOT2 Mouse Polyclonal Antibody
Catalog # ABNOH00004848-A01
Supplier:  Abnova
undefined
Anti-CNOT2 Mouse Polyclonal Antibody
Catalog # ABNOH00004848-A01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CCR4-NOT transcription complex, subunit 2
  • Antigen symbol:
    CNOT2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    4848
  • Antigen synonyms:
    HSPC131|CDC36|NOT2H|NOT2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: CNOT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human