Anti-NQO2 Rabbit Polyclonal Antibody
ABNOH00004835-D01P
: Abnova
- Antibody type:Primary
- Antigen name:NAD(P)H dehydrogenase, quinone 2
- Antigen symbol:NQO2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4835
- Antigen synonyms:QR2|NMOR2|DIA6|DHQV
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: NQO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human