Anti-MPZ Mouse Monoclonal Antibody [clone: 3B12]
Catalog # ABNOH00004359-M05
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Myelin Protein Zero
- Antigen symbol:MPZ
- Clonality:Monoclonal
- Clone:3B12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4359
- Antigen synonyms:P0|myelin protein zero (Charcot-Marie-Tooth neuropathy 1B) |CHM|CMT1B|CMT4E|MPZ|HMSNIB|CMT2I|myelin protein zero|MPP|CMTDI3|CMT2J|DSS|CMT1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MPZ
Clonality: Monoclonal
Clone: 3B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human