Anti-MAP3K4 Mouse Monoclonal Antibody [clone: 4C3]
Catalog # ABNOH00004216-M05
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase Kinase Kinase 4
- Antigen symbol:MAP3K4
- Clonality:Monoclonal
- Clone:4C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4216
- Antigen synonyms:MTK1|MEKK4|PRO0412|MEKK 4|MAPKKK4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MAP3K4
Clonality: Monoclonal
Clone: 4C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse