Anti-MAGEA4 Rabbit Polyclonal Antibody
Catalog # ABNOH00004103-D01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Melanoma Antigen Family A4
- Antigen symbol:MAGEA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4103
- Antigen synonyms:MAGE4A|CT1.4|MAGE4B|MAGE-X2|MAGE-41|MAGE4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: MAGEA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse