Anti-CK34 Mouse Monoclonal Antibody [clone: 3E5]
Catalog # ABNOH00003885-M10
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 34
- Antigen symbol:CK34
- Clonality:Monoclonal
- Clone:3E5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3885
- Antigen synonyms:KRTHA4|Ha-4|HA4|hHa4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CK34
Clonality: Monoclonal
Clone: 3E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human