Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-ANOS1 Mouse Monoclonal Antibody [clone: 2E8]
  ABNOH00003730-M01
 :  Abnova
undefined
Anti-ANOS1 Mouse Monoclonal Antibody [clone: 2E8]
  ABNOH00003730-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Anosmin 1
  • Antigen symbol:
    ANOS1
  • Clonality:
    Monoclonal
  • Clone:
    2E8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    3730
  • Antigen synonyms:
    HH1|KMS|KAL|ADMLX|HHA|WFDC19|KAL1|KALIG-1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: ANOS1
Clonality: Monoclonal
Clone: 2E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human