Anti-ITGB5 Rabbit Polyclonal Antibody
ABNOH00003693-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Integrin Beta 5
- Antigen symbol:ITGB5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3693
- Antigen synonyms:RGD1563276
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ITGB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse