Anti-HOXD1 Mouse Polyclonal Antibody
ABNOH00003231-B01
: Abnova
- Antibody type:Primary
- Antigen name:Homeobox D1
- Antigen symbol:HOXD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3231
- Antigen synonyms:Hox-4.7|HOX4G|HOX4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: HOXD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human