Anti-HELLS Mouse Monoclonal Antibody [clone: 2G10]
ABNOH00003070-M04
: Abnova
- Antibody type:Primary
- Antigen name:helicase, lymphoid-specific
- Antigen symbol:HELLS
- Clonality:Monoclonal
- Clone:2G10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3070
- Antigen synonyms:SMARCA6|LSH|Nbla10143|PASG
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: HELLS
Clonality: Monoclonal
Clone: 2G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human