Anti-GSTT1 Mouse Monoclonal Antibody [clone: 2D12-1C9]
ABNOH00002952-M02
: Abnova
- Antibody type:Primary
- Antigen name:Glutathione S-transferase theta 1
- Antigen symbol:GSTT1
- Clonality:Monoclonal
- Clone:2D12-1C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2952
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: GSTT1
Clonality: Monoclonal
Clone: 2D12-1C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human