Anti-GRN Rabbit Polyclonal Antibody
ABNOH00002896-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Granulin
- Antigen symbol:GRN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2896
- Antigen synonyms:proepithelin|PGRN|PC cell-derived growth factor|GEP|PCDGF|progranulin|granulin|PEPI|GRN|GP88|acrogranin|CLN11|granulin-epithelin
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: GRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse