Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-GNGT2 Mouse Monoclonal Antibody [clone: 1G11-C7]
  ABNOH00002793-M01
 :  Abnova
undefined
Anti-GNGT2 Mouse Monoclonal Antibody [clone: 1G11-C7]
  ABNOH00002793-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    G Protein Subunit Gamma Transducin 2
  • Antigen symbol:
    GNGT2
  • Clonality:
    Monoclonal
  • Clone:
    1G11-C7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    2793
  • Antigen synonyms:
    GNG8|GNG9|GNGT8|G-GAMMA-C|G-GAMMA-8
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: GNGT2
Clonality: Monoclonal
Clone: 1G11-C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human