Anti-GLS Mouse Monoclonal Antibody [clone: 5C4]
Catalog # ABNOH00002744-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Glutaminase
- Antigen symbol:GLS
- Clonality:Monoclonal
- Clone:5C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2744
- Antigen synonyms:GAM|KGA|GLS1|GAC|AAD20
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GLS
Clonality: Monoclonal
Clone: 5C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human