Order Entry
United Kingdom
ContactUsLinkComponent
Anti-EN1 Mouse Monoclonal Antibody [clone: 3H2]
Anti-EN1 Mouse Monoclonal Antibody [clone: 3H2]
Catalog # ABNOH00002019-M04
Supplier:  Abnova
undefined
Anti-EN1 Mouse Monoclonal Antibody [clone: 3H2]
Catalog # ABNOH00002019-M04
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Engrailed 1
  • Antigen symbol:
    EN1
  • Clonality:
    Monoclonal
  • Clone:
    3H2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    2019
  • Antigen synonyms:
    Homeobox protein engrailed-1|Hu-En-1|HME1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: EN1
Clonality: Monoclonal
Clone: 3H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human