Anti-PHC2 Mouse Polyclonal Antibody
Catalog # ABNOH00001912-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Polyhomeotic Homolog 2
- Antigen symbol:PHC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1912
- Antigen synonyms:EDR2|PH2|HPH2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: PHC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human